accessibility menu, dialog, popup

UserName

CAS RN 58438-04-3

CH3CH3H3COOHN(S)OO

CAS RN 58438-04-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol): 314.36
Molecular Formula: C18H20NO4
InChi Key: URKWHOVNPHQQTM-HNNXBMFYSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)