CAS RN 5834-16-2
CAS RN 5834-16-2
IUPAC Name:
3-methylthiophene-2-carbaldehyde
Synonyms:
3-methylthiophene-2-carbaldehyde
Molecular Weight (g/mol):
126.17
Molecular Formula:
C6H6OS
InChi Key:
BSQKBHXYEKVKMN-UHFFFAOYSA-N
SMILES:
CC1=C(SC=C1)C=O
1
–
1
of
1
results
1
3-Methylthiophene-2-carboxaldehyde, tech. 85%
CAS: 5834-16-2 Molecular Formula: C6H6OS Molecular Weight (g/mol): 126.173 MDL Number: MFCD00005430 InChI Key: BSQKBHXYEKVKMN-UHFFFAOYSA-N Synonym: 3-methyl-2-thiophenecarboxaldehyde,3-methylthiophene-2-carboxaldehyde,2-formyl-3-methylthiophene,3-methylthiophene-2-aldehyde,3-methyl-2-thiophenecarbaldehyde,3-methyl-2-formylthiophene,3-methyl-2-thiophene carboxaldehyde,2-thiophenecarboxaldehyde, 3-methyl,thiophenecarboxaldehyde, methyl,3-methyl-thiophene-2-carbaldehyde PubChem CID: 79911 IUPAC Name: 3-methylthiophene-2-carbaldehyde SMILES: CC1=C(SC=C1)C=O