accessibility menu, dialog, popup

UserName

CAS RN 5824-40-8

IUPAC Name: triphenylmethanamine
Synonyms: triphenylmethanamine
Molecular Weight (g/mol): 259.35
Molecular Formula: C19H17N
InChi Key: BZVJOYBTLHNRDW-UHFFFAOYSA-N
SMILES: NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)