accessibility menu, dialog, popup

UserName

CAS RN 5820-22-4

H3CCH2O

CAS RN 5820-22-4

IUPAC Name: [(2-methylprop-2-en-1-yl)oxy]benzene
Synonyms: [(2-methylprop-2-en-1-yl)oxy]benzene
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: LECDNXOCIPRJNJ-UHFFFAOYSA-N
SMILES: CC(=C)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results