CAS RN 58023-19-1
CAS RN 58023-19-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Bis(2-chloroethyl)benzenesulfonamide, TRC
CAS: 58023-19-1 Molecular Formula: C10 H13 Cl2 N O2 S Molecular Weight (g/mol): 282.19 Synonym: NSC 240403; IUPAC Name: N,N-bis(2-chloroethyl)benzenesulfonamide SMILES: ClCCN(CCCl)S(=O)(=O)c1ccccc1