accessibility menu, dialog, popup

UserName

CH3H3CBr

CAS RN 576-22-7

IUPAC Name: 2-bromo-1,3-dimethylbenzene
Synonyms: 2-bromo-1,3-dimethylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: MYMYVYZLMUEVED-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1Br
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)