accessibility menu, dialog, popup

UserName

H3COOF

CAS RN 57486-67-6

IUPAC Name: methyl 2-(2-fluorophenyl)acetate
Synonyms: methyl 2-(2-fluorophenyl)acetate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: TWXVEZBPKFIMBB-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results