CAS RN 573714-78-0
CAS RN 573714-78-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2H-1,2,3-Triazole-4-carboxylic Acid, TRC
CAS: 573714-78-0 Molecular Formula: C3 H3 N3 O2 Molecular Weight (g/mol): 113.075 IUPAC Name: 1H-triazole-4-carboxylic acid SMILES: OC(=O)c1c[nH]nn1