accessibility menu, dialog, popup

UserName

CAS RN 57-96-5

OSOONN

CAS RN 57-96-5

IUPAC Name: 4-[2-(benzenesulfinyl)ethyl]-1,2-diphenylpyrazolidine-3,5-dione
Synonyms: sulfinpyrazone anturane anturan apo sulfinpyrazone sulfinpirazona nu sulfinpyrazone nu-sulfinpyrazone enturan apo-sulfinpyrazone enturen
Molecular Weight (g/mol): 404.48
Molecular Formula: C23H20N2O3S
InChi Key: MBGGBVCUIVRRBF-UHFFFAOYNA-N
SMILES: O=C1C(CCS(=O)C2=CC=CC=C2)C(=O)N(N1C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results