accessibility menu, dialog, popup

UserName

H3COOCH3NSN

CAS RN 56891-69-1

IUPAC Name: [3-cyano-4-(ethoxycarbonyl)-6-methylpyridin-2-yl]sulfanide
Synonyms: [3-cyano-4-(ethoxycarbonyl)-6-methylpyridin-2-yl]sulfanide
Molecular Weight (g/mol): 221.25
Molecular Formula: C10H9N2O2S
InChi Key: WBMMFDGNOQLZNF-UHFFFAOYSA-M
SMILES: CCOC(=O)C1=CC(C)=NC([S-])=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results