accessibility menu, dialog, popup

UserName

H3CHNCH3

CAS RN 568577-84-4

IUPAC Name: [(4-ethylphenyl)methyl](methyl)amine
Synonyms: [(4-ethylphenyl)methyl](methyl)amine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: JCSDSVXBTRWEJS-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CNC)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results