accessibility menu, dialog, popup

UserName

CAS RN 56799-97-4

CH3OSOONH

CAS RN 56799-97-4

IUPAC Name: 4-methoxy-N-(naphthalen-1-yl)benzene-1-sulfonamide
Synonyms: 4-methoxy-N-(naphthalen-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 313.37
Molecular Formula: C17H15NO3S
InChi Key: KBIAXNFZLJUXNY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)NC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results