CAS RN 56752-55-7
CAS RN 56752-55-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Ethoxyhexa-3,5-diene, TRC
CAS: 56752-55-7 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.2 Synonym: 5-Ethoxy-1,3-hexadiene IUPAC Name: (3E)-5-ethoxyhexa-1,3-diene SMILES: CCOC(C)\C=C\C=C