CAS RN 5661-42-7
CAS RN 5661-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac N-Acetyl Ephedrine, TRC
CAS: 5661-42-7 Molecular Formula: C12H17NO2 Molecular Weight (g/mol): 207.27 Synonym: (R*,S*)-N-(2-Hydroxy-1-methyl-2-phenylethyl)-N-methylacetamide,erythro-N-(β-Hydroxy-α-methylphenethyl)-N-methylacetamide,N-Acetylephedrine; IUPAC Name: N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylacetamide SMILES: OC([C@H](N(C)C(C)=O)C)C1=CC=CC=C1