accessibility menu, dialog, popup

UserName

CAS RN 56553-60-7

BOOOOOOH3CCH3H3CHNa

CAS RN 56553-60-7

IUPAC Name: sodium bis(acetyloxy)boranuidyl acetate
Synonyms: sodium triacetoxyborohydride
Molecular Weight (g/mol): 211.94
Molecular Formula: C6H10BNaO6
InChi Key: HHYFEYBWNZJVFQ-UHFFFAOYSA-N
SMILES: [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)