CAS RN 565-20-8
CAS RN 565-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Acetyl Dapsone, TRC
CAS: 565-20-8 Molecular Formula: C14 H14 N2 O3 S Molecular Weight (g/mol): 290.34 Synonym: N-Acetyldapsone,Acetanilide, 4'-sulfanilyl- (6CI,7CI,8CI),N-[4-[(4-Aminophenyl)sulfonyl]phenyl]acetamide,4-Acetamido-4'-aminodiphenyl sulfone,4-Acetamidophenyl 4'-aminophenyl sulfone,4-Acetylamino-4'-aminodiphenyl sulfone,4-Acetylamino-4'-aminophenyl sulfone,4-Amino-4'-acetamidodiphenyl sulfone,4'-Sulfanilylacetanilide,Acetyldapsone,MADDS,Monoacetyldapsone,N-Acetyldapsone,NSC 27184,Acetamide, N-[4-[(4-aminophenyl)sulfonyl]phenyl]- IUPAC Name: N-[4-(4-aminophenyl)sulfonylphenyl]acetamide SMILES: CC(=O)Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2