accessibility menu, dialog, popup

UserName

CAS RN 55851-37-1

SOOHNClNOO

CAS RN 55851-37-1

IUPAC Name: N-(4-chloro-3-nitrophenyl)benzenesulfonamide
Synonyms: N-(4-chloro-3-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 312.72
Molecular Formula: C12H9ClN2O4S
InChi Key: OAEWODJOQXHPGS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=CC(NS(=O)(=O)C2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results