CAS RN 5582-62-7
IUPAC Name:
trimethyl(prop-2-yn-1-yloxy)silane
Synonyms:
trimethyl(prop-2-yn-1-yloxy)silane
Molecular Weight (g/mol):
128.25
Molecular Formula:
C6H12OSi
InChi Key:
ZZRPJWCNCLSOLR-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OCC#C
1
–
3
of
3
results
1
(Propargyloxy)trimethylsilane, 97%
CAS: 5582-62-7 Molecular Formula: C6H12OSi Molecular Weight (g/mol): 128.246 MDL Number: MFCD00042844 InChI Key: ZZRPJWCNCLSOLR-UHFFFAOYSA-N Synonym: propargyloxytrimethylsilane,propargyloxy trimethylsilane,o-trimethylsilylpropargyl alcohol,silane, trimethyl 2-propynyloxy,trimethyl prop-2-yn-1-yloxy silane,3-trimethylsiloxy-1-propyne,trimethyl 2-propynyloxy silane,silane, trimethyl 2-propyn-1-yloxy,trimethyl prop-2-ynyloxy silane,3-trimethylsiloxypropyne PubChem CID: 79696 IUPAC Name: trimethyl(prop-2-ynoxy)silane SMILES: C[Si](C)(C)OCC#C