CAS RN 5578-42-7
CAS RN 5578-42-7
IUPAC Name:
dichloro(cyclohexyl)methylsilane
Synonyms:
dichloro(cyclohexyl)methylsilane
Molecular Weight (g/mol):
197.17
Molecular Formula:
C7H14Cl2Si
InChi Key:
YUYHCACQLHNZLS-UHFFFAOYSA-N
SMILES:
C[Si](Cl)(Cl)C1CCCCC1
1
–
1
of
1
results
1
Cyclohexylmethyldichlorosilane, 97+%, Thermo Scientific Chemicals
CAS: 5578-42-7 Molecular Formula: C7H14Cl2Si Molecular Weight (g/mol): 197.18 InChI Key: YUYHCACQLHNZLS-UHFFFAOYSA-N Synonym: methylcyclohexyldichlorosilane,silane, dichlorocyclohexylmethyl,dichloro cyclohexyl methyl silane,cyclohexylmethyldichlorosilane,cyclohexane, dichloromethylsilyl,methyl cyclohexyldichlorosilane,cyclohexyl methyl dichlorosilane,cyclohexyl-dichloro-methylsilane,cyclohexyl-methyl-dichlorosilane,dichloro cyclohexyl methylsilane PubChem CID: 79685 IUPAC Name: dichloro-cyclohexyl-methylsilane SMILES: C[Si](C1CCCCC1)(Cl)Cl