accessibility menu, dialog, popup

UserName

CAS RN 55745-74-9

IUPAC Name: 1-(2,3-dihydro-1-benzofuran-5-yl)methanamine
Synonyms: 1-(2,3-dihydro-1-benzofuran-5-yl)methanamine
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: WQXWNTPLZFVZNX-UHFFFAOYSA-N
SMILES: NCC1=CC2=C(OCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results