accessibility menu, dialog, popup

UserName

AlH3CCH3CH3OCH3CH3CH3OCH3CH3CH3O

CAS RN 556-91-2

IUPAC Name: aluminium(3+) tris(2-methylpropan-2-olate)
Synonyms: aluminium(3+) tris(2-methylpropan-2-olate)
Molecular Weight (g/mol): 246.33
Molecular Formula: C12H27AlO3
InChi Key: MDDPTCUZZASZIQ-UHFFFAOYSA-N
SMILES: [Al+3].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)