accessibility menu, dialog, popup

UserName

CAS RN 55488-18-1

FeOOH2COOH2COOH2C

CAS RN 55488-18-1

IUPAC Name: iron(3+) tris(prop-2-enoate)
Synonyms: iron(3+) triacrylate
Molecular Weight (g/mol): 269.01
Molecular Formula: C9H9FeO6
InChi Key: WNWBIDPJHFYYLM-UHFFFAOYSA-K
SMILES: [Fe+3].[O-]C(=O)C=C.[O-]C(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results