CAS RN 554-35-8
CAS RN 554-35-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Linamarin, TRC
CAS: 554-35-8 Molecular Formula: C10 H17 N O6 Molecular Weight (g/mol): 247.25 IUPAC Name: 2-methyl-2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxypropanenitrile SMILES: CC(C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)C#N