CAS RN 55320-65-5
CAS RN 55320-65-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Hexanoic-2,2-d2 Acid, TRC
CAS: 55320-65-5 Molecular Formula: C6 D2 H10 O2 Molecular Weight (g/mol): 118.171 Synonym: Hexanoic-2,2-d2 acid (9CI),2,2-Dideuteriohexanoic acid,Hexanoic acid-d2,Hexanoic-α,α-d2 acid,Hexanoic-2,2-D2 acid,Caproic-α-d2 acid IUPAC Name: 2,2-dideuteriohexanoic acid SMILES: [2H]C([2H])(CCCC)C(=O)O
2
Hexanoic-2,2-d2 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.