accessibility menu, dialog, popup

UserName

CAS RN 55304-75-1

FFFClNCl

CAS RN 55304-75-1

IUPAC Name: 2,6-dichloro-3-(trifluoromethyl)pyridine
Synonyms: 2,6-dichloro-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.98
Molecular Formula: C6H2Cl2F3N
InChi Key: UPWAAFFFSGQECJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)N=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results