CAS RN 553-97-9
IUPAC Name:
2-methylcyclohexa-2,5-diene-1,4-dione
Synonyms:
2-methyl-1,4-benzoquinone
Molecular Weight (g/mol):
122.12
Molecular Formula:
C7H6O2
InChi Key:
VTWDKFNVVLAELH-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)C=CC1=O
1
–
1
of
1
results
1
Methyl-p-benzoquinone, 98%
CAS: 553-97-9 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 MDL Number: MFCD00001603 InChI Key: VTWDKFNVVLAELH-UHFFFAOYSA-N Synonym: p-toluquinone,methyl-p-benzoquinone,2-methyl-1,4-benzoquinone,toluquinone,methyl-1,4-benzoquinone,2-methyl-p-benzoquinone,tolylquinone,2-methylbenzoquinone,2-methylquinone,methylbenzoquinone PubChem CID: 11122 IUPAC Name: 2-methylcyclohexa-2,5-diene-1,4-dione SMILES: CC1=CC(=O)C=CC1=O