accessibility menu, dialog, popup

UserName

CH3CH3Br

CAS RN 553-94-6

IUPAC Name: 2-bromo-1,4-dimethylbenzene
Synonyms: benzene, 2-bromo-1,4-dimethyl-
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: QXISTPDUYKNPLU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(Br)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results