accessibility menu, dialog, popup

UserName

ONH

CAS RN 553-03-7

IUPAC Name: 1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: TZOYXRMEFDYWDQ-UHFFFAOYSA-N
SMILES: O=C1CCC2=CC=CC=C2N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results