CAS RN 551-43-9
CAS RN 551-43-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isomannide Dinitrate, TRC
CAS: 551-43-9 Molecular Formula: C6 H8 N2 O8 Molecular Weight (g/mol): 236.14 Synonym: Isomannit Dinitrate IUPAC Name: [(3R,3aS,6R,6aS)-6-nitrooxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3-yl] nitrate SMILES: [O-][N+](=O)O[C@@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+](=O)[O-]