accessibility menu, dialog, popup

UserName

CAS RN 54925-64-3

CH3H3CCH3SiH3CCH3NN

CAS RN 54925-64-3

IUPAC Name: 1-(tert-butyldimethylsilyl)-1H-imidazole
Synonyms: 1-(tert-butyldimethylsilyl)imidazole
Molecular Weight (g/mol): 182.34
Molecular Formula: C9H18N2Si
InChi Key: VUENSYJCBOSTCS-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)N1C=CN=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)