accessibility menu, dialog, popup

UserName

CH3NH2Br

CAS RN 54879-20-8

IUPAC Name: 2-bromo-3-methylaniline
Synonyms: 2-bromo-3-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: VJNUZLYTGSGDHR-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(N)=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results