accessibility menu, dialog, popup

UserName

CAS RN 5469-26-1

CH3CH3CH3OBr

CAS RN 5469-26-1

IUPAC Name: 1-bromo-3,3-dimethylbutan-2-one
Synonyms: 1-bromopinacolone
Molecular Weight (g/mol): 179.06
Molecular Formula: C6H11BrO
InChi Key: SAIRZMWXVJEBMO-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CBr
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)