accessibility menu, dialog, popup

UserName

CAS RN 5464-12-0

CH3NNOH

CAS RN 5464-12-0

IUPAC Name: 2-(4-methylpiperazin-1-yl)ethan-1-ol
Synonyms: 2-(4-methylpiperazin-1-yl)ethanol
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: QHTUMQYGZQYEOZ-UHFFFAOYSA-N
SMILES: CN1CCN(CCO)CC1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)