accessibility menu, dialog, popup

UserName

NNNHH2N

CAS RN 54608-52-5

IUPAC Name: 2-hydrazinylpyrazine
Synonyms: 2-hydrazinylpyrazine
Molecular Weight (g/mol): 110.12
Molecular Formula: C4H6N4
InChi Key: IVRLZJDPKUSDCF-UHFFFAOYSA-N
SMILES: NNC1=NC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results