accessibility menu, dialog, popup

UserName

CAS RN 54571-66-3

CH3OOOHN

CAS RN 54571-66-3

IUPAC Name: methyl 5-oxopyrrolidine-2-carboxylate
Synonyms: methyl 5-oxopyrrolidine-2-carboxylate
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: HQGPKMSGXAUKHT-UHFFFAOYNA-N
SMILES: COC(=O)C1CCC(=O)N1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)