accessibility menu, dialog, popup

UserName

CAS RN 5418-51-9

ONONHO

CAS RN 5418-51-9

IUPAC Name: 5-nitro-1,2-dihydropyridin-2-one
Synonyms: 5-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: XKWSQIMYNVLGBO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)