accessibility menu, dialog, popup

UserName

CAS RN 54-86-4

NaOON

CAS RN 54-86-4

IUPAC Name: sodium pyridine-3-carboxylate
Synonyms: sodium nicotinate
Molecular Weight (g/mol): 145.09
Molecular Formula: C6H4NNaO2
InChi Key: KFLRWGSAMLBHBV-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results