accessibility menu, dialog, popup

UserName

CAS RN 5391-30-0

NH2SNHBr

CAS RN 5391-30-0

IUPAC Name: (2-bromophenyl)thiourea
Synonyms: 2-bromophenylthiourea
Molecular Weight (g/mol): 231.11
Molecular Formula: C7H7BrN2S
InChi Key: QIGMVYSPXPXCPN-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results