accessibility menu, dialog, popup

UserName

CAS RN 53823-60-2

NaNaPdClClClClClCl

CAS RN 53823-60-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 365.1
Molecular Formula: Cl6Na2Pd
InChi Key: GTKIKTITWWGAJG-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[Pd]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)