accessibility menu, dialog, popup

UserName

CAS RN 5380-87-0

OOO

CAS RN 5380-87-0

IUPAC Name: 2-{[(oxiran-2-yl)methoxy]methyl}furan
Synonyms: 2-[(oxiran-2-ylmethoxy)methyl]furan
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: RUGWIVARLJMKDM-UHFFFAOYNA-N
SMILES: C(OCC1=CC=CO1)C1CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)