CAS RN 53558-93-3
CAS RN 53558-93-3
IUPAC Name:
(2R)-5-oxooxolane-2-carboxylate
Synonyms:
(2R)-5-oxooxolane-2-carboxylate
Molecular Weight (g/mol):
129.09
Molecular Formula:
C5H5O4
InChi Key:
QVADRSWDTZDDGR-GSVOUGTGSA-M
SMILES:
[O-]C(=O)[C@H]1CCC(=O)O1
1
–
3
of
3
results
1
(R)-5-Oxotetrahydrofuran-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-5-Oxotetrahydrofuran-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-5-Oxotetrahydrofuran-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.