accessibility menu, dialog, popup

UserName

CAS RN 53518-15-3

H2NOOFFF

CAS RN 53518-15-3

IUPAC Name: 7-amino-4-(trifluoromethyl)-2H-chromen-2-one
Synonyms: coumarin 151
Molecular Weight (g/mol): 229.16
Molecular Formula: C10H6F3NO2
InChi Key: JBNOVHJXQSHGRL-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results