accessibility menu, dialog, popup

UserName

CAS RN 5334-30-5

NH2NNNN

CAS RN 5334-30-5

IUPAC Name: 1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 1-phenylpyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 211.23
Molecular Formula: C11H9N5
InChi Key: KKDPIZPUTYIBFX-UHFFFAOYSA-N
SMILES: NC1=C2C=NN(C2=NC=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results