accessibility menu, dialog, popup

UserName

NaOONHO

CAS RN 532-94-5

IUPAC Name: sodium 2-(phenylformamido)acetate
Synonyms: sodium hippurate
Molecular Weight (g/mol): 201.16
Molecular Formula: C9H8NNaO3
InChi Key: ZBCAZEFVTIBZJS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CNC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results