accessibility menu, dialog, popup

UserName

CAS RN 532-91-2

CH3OHNOO

CAS RN 532-91-2

IUPAC Name: 6-methoxy-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-methoxy-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: MKMCJLMBVKHUMS-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=O)OC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results