CAS RN 5311-05-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methoxy-6-(trifluoromethyl)-1,3,5-triazin-2-amine, TRC
CAS: 5311-05-7 Molecular Formula: C5 H5 F3 N4 O Molecular Weight (g/mol): 194.11 Synonym: s-Triazine, 2-amino-4-methoxy-6-(trifluoromethyl)- (7CI,8CI),4-Methoxy-6-(trifluoromethyl)-1,3,5-triazin-2-amine,2-Amino-4-methoxy-6-(trifluoromethyl)-1,3,5-triazine IUPAC Name: 4-methoxy-6-(trifluoromethyl)-1,3,5-triazin-2-amine SMILES: COc1nc(N)nc(n1)C(F)(F)F