accessibility menu, dialog, popup

UserName

CAS RN 53104-95-3

OOFFFOH

CAS RN 53104-95-3

IUPAC Name: 4-hydroxy-3-(trifluoromethoxy)benzaldehyde
Synonyms: 4-hydroxy-3-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: GPJSLRQIOKFRFL-UHFFFAOYSA-N
SMILES: OC1=C(OC(F)(F)F)C=C(C=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results