accessibility menu, dialog, popup

UserName

CAS RN 531-75-9

IUPAC Name: 7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Synonyms: (-)-esculin aesculetin glukosid 7-hydroxy-6-cumarinyl-glucosid esculoside aesculin venoplant
Molecular Weight (g/mol): 340.28
Molecular Formula: C15H16O9
InChi Key: XHCADAYNFIFUHF-TVKJYDDYSA-N
SMILES: OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)