accessibility menu, dialog, popup

UserName

CAS RN 5307-99-3

ClClO

CAS RN 5307-99-3

IUPAC Name: 7,7-dichlorobicyclo[3.2.0]hept-2-en-6-one
Synonyms: 7,7-dichlorobicyclo[3.2.0]hept-2-en-6-one
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: JBPBARAOHIDZPU-UHFFFAOYNA-N
SMILES: ClC1(Cl)C2C=CCC2C1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results