accessibility menu, dialog, popup

UserName

CH3NNCH3BrBr

CAS RN 525596-66-1

IUPAC Name: 3-methyl-1-[12-(3-methylpyridin-1-ium-1-yl)dodecyl]pyridin-1-ium dibromide
Synonyms: 3-methyl-1-[12-(3-methylpyridin-1-ium-1-yl)dodecyl]pyridin-1-ium dibromide
Molecular Weight (g/mol): 514.39
Molecular Formula: C24H38Br2N2
InChi Key: NNPBJCAIRCFEFZ-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CC1=C[N+](CCCCCCCCCCCC[N+]2=CC=CC(C)=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)